Learn More
MINA53 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Marca:  Invitrogen PA511157
Detalles adicionales : Peso : 0.01000kg
Código de producto. 10372600
Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
Descripción
Descripción
MINA protein is directly involved in ribosome biogenesis, most likely during the assembly process of preribosomal particles. This protein is also involved in cell proliferation. MINA may have a role in esophageal squamous cell carcinoma, colon cancer and lung cancer.
Especificaciones
Especificaciones
MINA53
Polyclonal
RIOX2
Protein mina53; MYC-induced nuclear antigen; Mineral dust-induced gene protein 2; MINA; MDIG; mina53;
Rabbit
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
84864
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Immunohistochemistry, Western Blot
Unconjugated
Q8IUF8
RIOX2
KLH conjugated synthetic peptide between 397-429 amino acids from the C-terminal region of human MINA
400 μL