missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSIP1 Monoclonal Antibody (E.493.5), Invitrogen™
Rabbit Monoclonal Antibody
Marca: Invitrogen MA514821
Detalles adicionales : Peso : 0.01000kg
Descripción
It is not recommended to aliquot this antibody.
PSIP1 Monoclonal antibody specifically detects PSIP1 in Human, Mouse, Rat samples. It is validated for ChIP Assay, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Especificaciones
PSIP1 | |
Monoclonal | |
9 μg/mL | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and <0.02% sodium azide; pH 7.5 | |
O75475, Q812D1, Q99JF8 | |
PSIP1 | |
Synthetic peptide corresponding to amino acids surrounding Lys461 of human LEDGF | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
ChIP Assay, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
E.493.5 | |
Unconjugated | |
PSIP1 | |
AA408851; AU015605; CLL-associated antigen KW-7; dense fine speckles 70; dense fine speckles 70 kDa protein; DFS 70; DFS70; LEDGF; Ledgfa; Ledgfb; Lens epithelium-derived growth factor; mLEDGF; p52; p75; PAIP; PC4 and SFRS1 interacting protein 1; PC4 and SFRS1 interacting protein 2; PC4 and SFRS1-interacting protein; PSIP1; Psip2; SBP75; supercoiled DNA binding protein 75; transcriptional coactivator p52/p75; Transcriptional coactivator p75/p52 | |
Rabbit | |
Affinity chromatography | |
RUO | |
101739, 11168, 313323 | |
-20°C | |
Liquid |