missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLF8 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA540390
Detalles adicionales : Peso : 0.00010kg
Descripción
Peptide sequence: LSFHKPKAPL QPASMLQAPI RPPKPQSSPQ TLVVSTSTSD MSTSANIPTV Sequence homology: Cow: 79%; Dog: 79%; Horse: 77%; Human: 100%; Rabbit: 86%
KLF8 Polyclonal antibody specifically detects KLF8 in Human samples. It is validated for Immunohistochemistry, Western Blot.Especificaciones
KLF8 | |
Polyclonal | |
Unconjugated | |
KLF8 | |
A830097P10Rik; Basic krueppel-like factor 3; basic kruppel-like factor 3; BKLF3; DKFZp686O08126; DXS741; KLF8; Klf8-ps1; Krueppel-like factor 8; Kruppel like factor 8; Kruppel-like factor 8; Kruppel-like factor 8, pseudogene 1; LOW QUALITY PROTEIN: Krueppel-like factor 8; MGC138314; RGD1565831; RP13-1021K9.1; zinc finger protein 741; ZNF74; ZNF741 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
11279 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
O95600 | |
KLF8 | |
Synthetic peptide directed towards the N-terminal of human KLF8 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |