missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IDNK Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA546105
Detalles adicionales : Peso : 0.00010kg
Descripción
Peptide sequence: RVVLACSALK KTYRDILTQG KDGVALKCEE SGKEAKQAEM QLLVVHLSGS Sequence homology: Human: 100%
IDNK Polyclonal antibody specifically detects IDNK in Human samples. It is validated for Western Blot.Especificaciones
IDNK | |
Polyclonal | |
Unconjugated | |
Idnk | |
5133401N09Rik; bA522I20.2; BCRG2; C9orf103; glucokinase-like protein; gluconate kinase; gluconokinase-like protein; hGntK; Idnk; idnK gluconokinase homolog (E. coli); IDNK, gluconokinase; idnK, gluconokinase homolog; KIAA1824; probable gluconokinase; WDR22 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
414328 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q5T6J7 | |
Idnk | |
Synthetic peptide directed towards the middle region of human IDNK | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |