missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GTF2A1 Monoclonal Antibody (2H5), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA519032
Detalles adicionales : Peso : 0.00010kg
Description
Peptide Sequence: MANSANTNTV PKLYRSVIED VINDVRDIFL DDGVDEQVLM ELKTLWENKL MQSRAVDGFH SEE
Transcription initiation factor IIA alpha; and beta; chains (GTF2A1, TFIIA p35 and p19 subunits, TFIIAL) binding ability of TLP is required for characteristic cytoplasmic localization of TLP. TFIIA may regulate the intracellular molecular state and the function of TLP through its property of binding to TLP.Specifications
GTF2A1 | |
Monoclonal | |
0.2-1.0 mg/mL | |
PBS with no preservative; pH 7.4 | |
P52655 | |
GTF2A1 | |
GTF2A1 (NP_056943, 1 a.a. ∽ 63 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
ELISA, Western Blot | |
2H5 | |
Unconjugated | |
GTF2A1 | |
TF2A1, TFIIA, TFIIA alpha, p55, TFIIA-42, TFIIAL, general transcription factor IIA subunit 1, glucose regulated protein, 58kD pseudogene, transcription initiation factor IIA subunit 1, transcription initiation factor TFIIA 42 kDa subunit | |
Mouse | |
Affinity chromatography | |
RUO | |
2957 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |