missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Goat IgG (H+L) Cross-Adsorbed Secondary Antibody, Biotin-XX, Invitrogen™
Rabbit Polyclonal Secondary Antibody
Marca: Invitrogen A10518
Detalles adicionales : Peso : 0.00450kg
Descripción
Goat IgG (H+L) Cross-Adsorbed Polyclonal antibody specifically detects Goat IgG (H+L) Cross-Adsorbed in Goat samples. It is validated for Immunocytochemistry.Especificaciones
Goat IgG (H+L) Cross-Adsorbed | |
Polyclonal | |
Biotin-XX | |
Rabbit | |
purified | |
RUO | |
Goat | |
Liquid |
Immunocytochemistry | |
2 mg/mL | |
PBS with 5mM sodium azide; pH 7.5 | |
Gamma Immunoglobins Heavy and Light chains | |
1 mg | |
Secondary | |
4° C | |
IgG |