missing translation for 'onlineSavingsMsg'
Learn More

GCN5 Monoclonal Antibody (S.775.4), Invitrogen™

Rabbit Monoclonal Antibody

Marca:  Invitrogen MA514884

Detalles adicionales : Peso : 0.01000kg

Código de producto. 11564851

  • 467.00€ / 100 microlitros
Fecha estimada de envío: 15-05-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

It is not recommended to aliquot this antibody. This antibody is not cross-reactive with the related PCAF protein.

GCN5 Monoclonal antibody specifically detects GCN5 in Human, Mouse, Non-human Primate, Rat samples. It is validated for Immunocytochemistry, Immunoprecipitation, Western Blot.
Especificaciones

Especificaciones

GCN5
Monoclonal
134 μg/mL
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and <0.02% sodium azide; pH 7.5
Q92830, Q9JHD2
KAT2A
Synthetic peptide corresponding to the amino terminus of human GCN5L2
100 μL
Primary
Human, Mouse, Non-human Primate, Rat
Antibody
IgG
Immunocytochemistry, Immunoprecipitation, Western Blot
S.775.4
Unconjugated
KAT2A
1110051E14Rik; AAS104; ADA4; AW212720; GCN5; GCN5 (general control of amino-acid synthesis, yeast, homolog)-like 2; GCN5 general control of amino acid synthesis-like 2; Gcn5l2; general control of amino acid synthesis 5-like 2; general control of amino acid synthesis protein 5-like 2; general control of amino acid synthesis, yeast homolog-like 2; General control of amino acid synthesis, yeast, homolog-like 2; general control of amino acid synthesis-like 2; hGCN5; Histone acetyltransferase GCN5; histone acetyltransferase KAT2A; Histone succinyltransferase KAT2A; hsGCN5; K(lysine) acetyltransferase 2A; KAT2; Kat2a; KATA2; Lysine acetyltransferase 2A; MmGCN5; PCAF-b; PCAF-B/GCN5; STAF97; SWI9; YGR252W
Rabbit
Affinity chromatography
RUO
14534, 2648, 303539
-20°C
Liquid
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales