missing translation for 'onlineSavingsMsg'
Learn More
Learn More
mGluR3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA129424
Detalles adicionales : Peso : 0.01000kg
Descripción
Recommended positive controls: Rat Cerebellar and cortical Membrane.
mGluR3 Polyclonal antibody specifically detects mGluR3 in Human, Mouse, Rat samples. It is validated for Western Blot.Especificaciones
mGluR3 | |
Polyclonal | |
tris glycine with 1% BSA and 0.1% sodium azide; pH 7.8 | |
P31422, Q14832, Q9QYS2 | |
GRM3 | |
synthetic peptide TKVDEAEYM, corresponding to amino acids 118-126 of Rat metabotropic Glutamate Receptor 3. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
GRM3 | |
0710001G23Rik; G protein-coupled receptor, family C, group 1, member C; GLUR3; glutamate metabotropic receptor 3; glutamate receptor; glutamate receptor, metabotropic 3; Gprc1c; GRM3; M mGlu3; Metabotropic glutamate receptor 3; mGlu3; mGluR3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
108069, 24416, 2913 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |