missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FSTL1 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marca: Invitrogen PA518086
Detalles adicionales : Peso : 0.01000kg
Descripción
This antibody is predicted to react with bovine based on sequence homology.
FSTL1 Polyclonal antibody specifically detects FSTL1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot.Especificaciones
FSTL1 | |
Polyclonal | |
Unconjugated | |
Fstl1 | |
AI316791; AW107808; FLJ50214; FLJ52277; follistatin like 1; follistatin-like 1; follistatin-like protein 1; Follistatin-related protein 1; FRP; FSL1; Fstl; FSTL1; MIR198; OCC1; OCC-1; TGF-beta-inducible protein TSC-36; tsc36; TSC-36 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
11167 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunocytochemistry, Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q12841 | |
Fstl1 | |
Synthetic peptide sequence (TAEKTKRVSTKEI) corresponding to the C-terminus amino acids of FSTL1 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |