missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGFR1 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marca: Invitrogen PA518344
Detalles adicionales : Peso : 0.01000kg
Descripción
This antibody is predicted to react with bovine and canine based on sequence homology.
FGFR1 Polyclonal antibody specifically detects FGFR1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Especificaciones
FGFR1 | |
Polyclonal | |
Unconjugated | |
FGFR1 | |
2.7.10.1; AW208770; Basic fibroblast growth factor receptor 1; basic fibroblast growth factor receptor I; BFGFR; bFGF-R; bFGF-R-1; CD331; CD331 antigen; CEK; CEK1; cek1 protein; cek1 protein precursor; c-fgr; Eask; ECCL; FGF receptor; FGF receptor-1; FGFBR; fgfr 1; Fgfr1; FGFR-1; FGFR1/PLAG1 fusion; FGFR1IIIC; FGFRI; Fibroblast growth factor receptor 1; fibroblast growth factor receptor 1 (fms-related tyrosine kinase 2, Pfeiffer syndrome); fibroblast growth factor receptor 1-IIIc; fibroblast growth factor receptor type 1; Flg; FLT2; FLT-2; FMS-like tyrosine kinase 2; fms-related tyrosine kinase 2; HBGFR; heparin-binding growth factor receptor; HH2; HRTFDS; Hspy; hydroxyaryl-protein kinase; KAL2; LOW QUALITY PROTEIN: fibroblast growth factor receptor 1; MFR; N-sam; OGD; proto-oncogene c-Fgr; Tyrosine kinase receptor CEK1 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
2260 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P11362 | |
FGFR1 | |
Synthetic peptide sequence (CLPRHPAQLANGGLKR) corresponding to the C-terminus amino acids of FGFR1 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |