missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DYKDDDDK Tag Monoclonal Antibody (FG4R), DyLight™ 488, Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA191878D488
Detalles adicionales : Peso : 0.01000kg
Descripción
MA1-91878-D488 has been successfully used for immunofluorescence. DyLight 488 has an excitation/emission of 493/518 nm. similar epitope as the FLAG tag from Sigma
DYKDDDDK Tag Monoclonal antibody specifically detects DYKDDDDK Tag in Tag samples. It is validated for Immunocytochemistry, Western Blot.Especificaciones
DYKDDDDK Tag | |
Monoclonal | |
1 mg/mL | |
PBS with proprietary stabilizer and 0.02% sodium azide | |
Mouse | |
Protein A | |
RUO | |
4° C, do not freeze | |
Liquid | |
Tag |
Immunocytochemistry, Western Blot | |
FG4R | |
DyLight 488 | |
DDDDK; DDDDK epitope tag; ECS epitope tag; ECS tag; FLAG; FLAG Tag | |
Synthetic peptide (DYKDDDDK) coupled to KLH. | |
50 μL | |
Primary | |
Antibody | |
IgG2b |