missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DND1 Monoclonal Antibody (1F7), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA523282
Detalles adicionales : Peso : 0.00010kg
Descripción
Peptide Sequence: EARLLPSPGP APGQIALLKF SSHRAAAMAK KALVEGQSHL CGEQVAVEWL KPDLKQRLRQ QLVGPFLRSP QPEGSQLALA RDKLGFQGAR ATLQ
DND1 Monoclonal antibody specifically detects DND1 in Human samples. It is validated for ELISA, Immunocytochemistry, Western Blot.Especificaciones
DND1 | |
Monoclonal | |
1.0 mg/mL | |
PBS with no preservative; pH 7.4 | |
Q8IYX4 | |
DND1 | |
DND1 (NP_919225, 167 a.a. ∽ 260 a.a) full length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
ELISA, Immunocytochemistry, Western Blot | |
1F7 | |
Unconjugated | |
DND1 | |
BC034897; dead end homolog 1; dead end homolog 1 (zebrafish); dead end protein homolog 1; DND microRNA-mediated repression inhibitor 1; Dnd1; MGC34750; Rbms4; RNA-binding motif, single-stranded-interacting protein 4; Ter; teratoma | |
Mouse | |
Affinity chromatography | |
RUO | |
373863 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |