missing translation for 'onlineSavingsMsg'
Learn More

CRIM1 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marca:  Invitrogen PA534410

Detalles adicionales : Peso : 0.01000kg

Código de producto. 15325341

  • 473.00€ / 100 microgramos
Fecha estimada de envío: 23-04-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

A suggested positive control is Jurkat cell lysate. PA5-34410 can be used with blocking peptide PEP-1452.

CRIM1 Polyclonal antibody specifically detects CRIM1 in Human samples. It is validated for Western Blot.
Especificaciones

Especificaciones

CRIM1
Polyclonal
Unconjugated
CRIM1
AU015004; Crim1; CRIM-1; cysteine rich transmembrane BMP regulator 1; cysteine rich transmembrane BMP regulator 1 (chordin like); cysteine rich transmembrane BMP regulator 1 (chordin-like); cysteine-rich motor neuron 1; cysteine-rich motor neuron 1 protein; Cysteine-rich repeat-containing protein S52; Processed cysteine-rich motor neuron 1 protein; S52; UNQ1886/PRO4330
Rabbit
Antigen affinity chromatography
RUO
51232
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Liquid
Western Blot
1 mg/mL
PBS with 0.02% sodium azide
Q9NZV1
CRIM1
A 17 amino acid synthetic peptide near the carboxy terminus of human Crim1
100 μg
Primary
Human
Antibody
IgG
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales