Learn More
TPL2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA521650
Detalles adicionales : Peso : 0.01000kg
Descripción
Recommended positive controls: 293T, A431, HeLa, HepG2, Raji, mouse liver. Predicted reactivity: Mouse (80%), Pig (80%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
TPL2 Polyclonal antibody specifically detects TPL2 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Especificaciones
TPL2 | |
Polyclonal | |
Unconjugated | |
MAP3K8 | |
augmented in rheumatoid arthritis 2; AURA2; Cancer Osaka thyroid oncogene; cancer Osaka thyroid, oncogene; CCOT; c-COT; COT; cot (cancer Osaka thyroid) oncogene; COT proto-oncogene serine/threonine-protein kinase; Cot/Tpl2; D17TPL2; EC 1.14.16.2; Est; Estf; Ewing sarcoma transformant; FLJ10486; M3K8; MAP3K 8; Map3k8; MEKK8; mitogen activated protein kinase kinase kinase 8; mitogen-activated protein kinase kinase kinase 8; Protein kinase cot; Proto-oncogene c-Cot; proto-oncogene serine/threoine protein kinase; serine/threonine kinase (Tpl-2); serine/threonine-protein kinase cot; TH isoform 3; TH isoform a; TH-4; TPL 2; Tpl2; TPL-2; Tumor progression locus 2; TY3H; TYH | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1326, 26410 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
P41279, Q07174 | |
MAP3K8 | |
Recombinant fragment corresponding to a region within amino acids 140 and 462 of CROT (Uniprot ID#Q9UKG9) | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |