Learn More
Complement Factor D Monoclonal Antibody (I8/1b), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen GAU0080102
Detalles adicionales : Peso : 0.01000kg
Descripción
GAU 008-01-02 detects Complement factor D in human serum and plasma. GAU 008-01-02 has been successfully used in ELISA and Western blot procedures. Use in Western blotting under non-reducing conditions. GAU 008-01 may be used as detection antibody in sandwich ELISA with GAU 010-04-02 as a capture antibody.
Complement Factor D Monoclonal antibody specifically detects Complement Factor D in Human samples. It is validated for ELISA, Western Blot.Especificaciones
Complement Factor D | |
Monoclonal | |
0.99 mg/mL | |
PBS with 15mM sodium azide; pH 7.4 | |
P00746 | |
CFD | |
Human complement factor D. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
ELISA, Western Blot | |
I8/1b | |
Unconjugated | |
CFD | |
28 kDa adipocyte protein; adipsin; Adn; C3 convertase activator; Cfd; Complement factor D; complement factor D (adipsin); complement factor D preproprotein; D component (adipsin) of complement; D component of complement (adipsin); Df; EC 3.4.21.46; Endogenous vascular elastase; EVE; PFD; Properdin factor D | |
Mouse | |
Protein A/G | |
RUO | |
1675 | |
4° C, store in dark | |
Liquid |