missing translation for 'onlineSavingsMsg'
Learn More

Cardiac Myosin Heavy Chain Monoclonal Antibody (3-48), Invitrogen™

Mouse Monoclonal Antibody

Marca:  Invitrogen MA126180

Detalles adicionales : Peso : 1.24740kg

Código de producto. 11559452

  • 516.00€ / 100 microgramos
Fecha estimada de envío: 23-04-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

This antibody is myosin alpha and beta heavy chain specific. Affinity constants: Human ventricular myosin heavy chains - 3.33 x 108M-1; Human atrial myosin heavy chains - 1.48 x 108M-1; Human skeletal muscle myosin - 1.06 x 108M-1; Canine cardiac myosin heavy chains - 2.06 x 108M-1. This antibody cross-reacts with heavy chains of human myosin isolated from slow skeletal muscle, fast skeletal muscle, eye muscle, and streptococcal M5 peptide. It does not react with human platelets, smooth muscle myosin heavy chains, myosin light chains, myosin fragments circulating in blood of patients with myocardial infarction, nor does it react with any other human tissue antigens or blood cells. Recommended positive control: Slow skeletal muscle, fast skeletal muscle, eye muscle.

Cardiac Myosin Heavy Chain Monoclonal antibody specifically detects Cardiac Myosin Heavy Chain in Bovine, Canine, Human, Mouse, Porcine, Rabbit, Rat samples. It is validated for ELISA, Immunocytochemistry, Immunohistochemistry (Frozen), Western Blot.
Especificaciones

Especificaciones

Cardiac Myosin Heavy Chain
Monoclonal
1 mg/mL
PBS with no preservative; pH 7.4
A2AQP0, A7E2Y1, P02563, P02564, P04460, P04461, P12883, P13533, P49824, P79293, Q02566, Q91Z83, Q9BE39
LOC100736765, MYH6, Myh7, Myh7b
Purified, full length, native human protein.
100 μg
Primary
Bovine, Canine, Human, Mouse, Porcine, Rabbit, Rat
Antibody
IgG1
ELISA, Immunocytochemistry, Immunohistochemistry (Frozen), Western Blot
3-48
Unconjugated
Myh7b
A830009F23Rik; AA517445; alpha cardiac MHC; alpha myosin; alpha-cardiac myosin heavy chain; alpha-MHC; alpha-myosin; alpha-myosin heavy chain; alpha-myosin heavy chain subfragment 2 exon; antigen MLAA-21; ASD3; beta cardiac myosin heavy chain; beta myosin heavy chain; beta-MHC; beta-myosin; beta-myosin heavy chain; B-MHC; cardiac muscle myosin heavy chain 7 beta; cardiac myosin heavy chain alpha isoform; cardiac myosin heavy chain beta isoform; cardiomyopathy, hypertrophic 1; CMD1EE; CMD1S; CMH1; CMH14; KIAA1512; LOC100736765; LOW QUALITY PROTEIN: myosin-6; LOW QUALITY PROTEIN: myosin-7B; MHC14; MPD1; Myh14; MYH6; MYH7; Myh7b; MYH-beta/slow; MYHC; Myhca; Myhc-a; MyHC-alpha; MYHCB; Myhc-b; MyHC-beta; MyHC-I; MyHC-slow; myopathy, distal 1; myosin alpha heavy chain; Myosin cardiac muscle beta chain; myosin heavy chain; myosin heavy chain 6; Myosin heavy chain 7; myosin heavy chain 7B; myosin heavy chain 7B, cardiac muscle beta isoform; myosin heavy chain polypeptide 6 cardiac muscle adult
Mouse
IgG fraction
RUO
100101566, 100125987, 100157870, 100296004, 100356178, 100685644, 100736765, 140781, 17888, 282714, 29556, 29557, 311570, 396860, 403807, 4624, 4625, 477206, 521764, 57644, 668940
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales