missing translation for 'onlineSavingsMsg'
Learn More

MRP3 Monoclonal Antibody (M3II-9), Invitrogen™

Mouse Monoclonal Antibody

Marca:  Invitrogen MA135680

Detalles adicionales : Peso : 0.01000kg

Código de producto. 11584982

  • 684.00€ / 1 ml
Fecha estimada de envío: 08-05-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

MRP3 Monoclonal antibody specifically detects MRP3 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Frozen), Western Blot.
Especificaciones

Especificaciones

MRP3
Monoclonal
Conc. Not Determined
tissue culture supernatant with 0.7% BSA and 0.1% sodium azide
O15438
ABCC3
ABCC3.
RUO
8714
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Liquid
Immunocytochemistry, Immunohistochemistry (Frozen), Western Blot
M3II-9
Unconjugated
ABCC3
ABC31; ABCC3; ATP binding cassette subfamily C member 3; ATP-binding cassette sub-family C member 3; ATP-binding cassette, subfamily C (CFTR/MRP), member 3; ATP-binding cassette, sub-family C (CFTR/MRP), member 3; canalicular multispecific organic anion transporter 2; canicular multispecific organic anion transporter; cMOAT2; DKFZp686E22157; EST90757; MLP2; MLP-2; MOAT-D; Mrp3; MRP-like protein 2; multidrug resistance associated protein; multidrug resistance protein 3; multidrug resistance-associated protein 3; multi-specific organic anion transporter D; organic anion transporter
Mouse
1 mL
Primary
Human
Antibody
IgG1
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales