Learn More
ALKBH8 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA558517
Detalles adicionales : Peso : 0.02000kg
Descripción
Immunogen sequence: LPPGLMVVEE IISSEEEKML LESVDWTEDT DNQNSQKSLK HRRVKHFGYE FHYENNNVDK DKPLSGGLPD ICESFLEKWL RKGYIKHKPD QMTINQYEPG QG Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 84%.
ALKBH8 Polyclonal antibody specifically detects ALKBH8 in Human samples. It is validated for Immunohistochemistry.Especificaciones
ALKBH8 | |
Polyclonal | |
Unconjugated | |
ALKBH8 | |
4930562C03Rik; 8030431D03Rik; 9430088N01Rik; ABH8; alkB homolog 8, tRNA methyltransferase; AlkB homologue 8; alkB, alkylation repair homolog 8; alkB, alkylation repair homolog 8 (E. coli); Alkbh8; Alkylated DNA repair protein alkB homolog 8; LOW QUALITY PROTEIN: alkylated DNA repair protein alkB homolog 8; probable alpha-ketoglutarate-dependent dioxygenase ABH8; RGD1304687; S-adenosyl-L-methionine-dependent tRNA methyltransferase ABH8; TRM9; TRMT9; tRNA (carboxymethyluridine(34)-5-O)-methyltransferase ABH8; tRNA methyltransferase 9 homolog | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
91801 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96BT7 | |
ALKBH8 | |
Recombinant protein corresponding to Human ALKBH8 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only