missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Agarosa, Grado de Tecnología Genética
Click to view available options
Cantidad:
25 g
100 g
Descripción
Engineered to permit recovery of active DNA after separation, with minimal inhibition of restriction and ligation enzymes
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Diseñado para permitir la recuperación de ADN activo después de la separación, con una inhibición mínima de las enzimas de restricción y ligación
Especificaciones
Nombre del producto químico o material | Agarose |
Forma física | Polvo |
Color | Blanco |
Cantidad | 25 g |
Grado | Tecnología genética |
Sensibilidad | Hygroscopic; Moisture sensitive |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'