Learn More
Fisherbrand™ Active Carbon Line Filter
Used for filtration of samples of hydrocarbons, cetone, alcohols, ether, organic acids, ambient air, various gases and smokes.
Marca: Fisherbrand™
Detalles adicionales : Peso : 0.01000kg
Descripción
- Composition of polycarbonate body with granules of coal and cellulose
- Can be used for temperatures up to 70°C
- 4 bar pressure resistant
Especificaciones
For filtration of samples of Hydrocarbons, Cetone, Alcohols, Ether, Organic Acids, ambient air, various gases, smokes | |
Filter | |
82 mm | |
26 mm |