missing translation for 'onlineSavingsMsg'
Learn More

Éster de pinacol de ácido 4-[(trimetilsilil)etinil]bencenoborónico, 97 %, Thermo Scientific™

Catalog Number p-7034049
Click to view available options
Cantidad:
1 g
5 g
Este artículo no se puede devolver. Vea la política de devoluciones
870238-65-6
C17H25BO2Si
300.28
MFCD08706286
CIKKODWPGJIJQG-UHFFFAOYSA-N
4,4,5,5-tetramethyl-2-4-trimethylsilanylethynyl-phenyl-1,3,2 dioxaborolane, 4-trimethylsilyl ethynyl phenylboronic acid pinacol ester, 1,3,2-dioxaborolane,4,4,5,5-tetramethyl-2-4-2-trimethylsilyl ethynyl phenyl, trimethyl 2-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl ethynyl silane, 4,4,5,5-tetramethyl-2-4-trimethylsilyl ethynyl phenyl-1,3,2-dioxaborolane, amtb200, 4-trimethylsilyl ethynyl benzeneboronic acid pinacol ester, 2-4-trimethylsilylethynyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
17999152
CC1(C)OB(OC1(C)C)C1=CC=C(C=C1)C#C[Si](C)(C)C
Este artículo no se puede devolver. Vea la política de devoluciones
870238-65-6
C17H25BO2Si
300.28
MFCD08706286
CIKKODWPGJIJQG-UHFFFAOYSA-N
4,4,5,5-tetramethyl-2-4-trimethylsilanylethynyl-phenyl-1,3,2 dioxaborolane, 4-trimethylsilyl ethynyl phenylboronic acid pinacol ester, 1,3,2-dioxaborolane,4,4,5,5-tetramethyl-2-4-2-trimethylsilyl ethynyl phenyl, trimethyl 2-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl ethynyl silane, 4,4,5,5-tetramethyl-2-4-trimethylsilyl ethynyl phenyl-1,3,2-dioxaborolane, amtb200, 4-trimethylsilyl ethynyl benzeneboronic acid pinacol ester, 2-4-trimethylsilylethynyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
17999152
CC1(C)OB(OC1(C)C)C1=CC=C(C=C1)C#C[Si](C)(C)C

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 4-[(Trimethylsilyl)ethynyl]benzeneboronic acid pinacol ester
Cantidad 1 g
Nombre IUPAC Trimetil({2-[4-(4,4,5,5-tetrametil-1,3,2-dioxaborolan-2-il)fenil]etynil})silano
Formula Weight (peso de la fórmula) 300.28
Porcentaje de pureza 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'