missing translation for 'onlineSavingsMsg'
Learn More

Ácido 4-formilfenilborónico, 97 %, Thermo Scientific Chemicals

Catalog Number p-2690679
Click to view available options
Cantidad:
1 g
5 g
25 g
Envase:
Frasco de vidrio
Este artículo no se puede devolver. Vea la política de devoluciones
87199-17-5
C7H7BO3
149.94
MFCD00151823
VXWBQOJISHAKKM-UHFFFAOYSA-N
4-formylphenyl boronic acid, 4-formylbenzeneboronic acid, 4-boronobenzaldehyde, boronic acid, 4-formylphenyl, boronic acid, b-4-formylphenyl, p-formylphenylboronic acid, 4-dihydroxyboryl benzaldehyde, 4-dihydroxyboranyl benzaldehyde, 4-benzaldehyde boronic acid, 4-formyl-phenylboronic acid
591073
(4-formylphenyl)boronic acid
OB(O)C1=CC=C(C=O)C=C1
Este artículo no se puede devolver. Vea la política de devoluciones
87199-17-5
C7H7BO3
149.94
MFCD00151823
VXWBQOJISHAKKM-UHFFFAOYSA-N
4-formylphenyl boronic acid, 4-formylbenzeneboronic acid, 4-boronobenzaldehyde, boronic acid, 4-formylphenyl, boronic acid, b-4-formylphenyl, p-formylphenylboronic acid, 4-dihydroxyboryl benzaldehyde, 4-dihydroxyboranyl benzaldehyde, 4-benzaldehyde boronic acid, 4-formyl-phenylboronic acid
591073
(4-formylphenyl)boronic acid
OB(O)C1=CC=C(C=O)C=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 4-Formylphenylboronic acid
Nombre de nota 97%
% mín. CAS 96.0
% máx. CAS 100.0
Punto de fusión 256°C to 257°C
Color Incoloro a naranja amarillento
Espectro infrarrojo Auténtico
Intervalo de porcentaje de ensayo 96% min. (HPLC)
Envase Frasco de vidrio
Cantidad 1 g
Formula Weight (peso de la fórmula) 149.94
Porcentaje de pureza 97%
Forma física Líquido
Mostrar más Mostrar menos

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'