missing translation for 'onlineSavingsMsg'
Learn More

Ácido 4-(dietilcarbamoil)bencenoborónico, 98 %, Thermo Scientific Chemicals

Catalog Number p-7035272
Click to view available options
Cantidad:
1 g
5 g
Este artículo no se puede devolver. Vea la política de devoluciones
389621-80-1
C11H16BNO3
221.06
MFCD03411949
ZCGVBHIMRVYWOH-UHFFFAOYSA-N
4-n,n-diethylaminocarbonyl phenylboronic acid, 4-diethylcarbamoyl phenylboronic acid, 4-diethylcarbamoyl phenyl boronic acid, 4-diethylcarbamoyl benzeneboronic acid, boronic acid, 4-diethylamino carbonyl phenyl, 4-diethylaminocarbonyl phenylboronic acid, acmc-209j2q, amtb302, n,n-diethyl 4-boronobenzamide, 4-diethylcarbamoyl benzeneboronic acid
2773375
ácido [4-(dietilcarbamoil)fenil]borónico
CCN(CC)C(=O)C1=CC=C(C=C1)B(O)O
Este artículo no se puede devolver. Vea la política de devoluciones
389621-80-1
C11H16BNO3
221.06
MFCD03411949
ZCGVBHIMRVYWOH-UHFFFAOYSA-N
4-n,n-diethylaminocarbonyl phenylboronic acid, 4-diethylcarbamoyl phenylboronic acid, 4-diethylcarbamoyl phenyl boronic acid, 4-diethylcarbamoyl benzeneboronic acid, boronic acid, 4-diethylamino carbonyl phenyl, 4-diethylaminocarbonyl phenylboronic acid, acmc-209j2q, amtb302, n,n-diethyl 4-boronobenzamide, 4-diethylcarbamoyl benzeneboronic acid
2773375
ácido [4-(dietilcarbamoil)fenil]borónico
CCN(CC)C(=O)C1=CC=C(C=C1)B(O)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 4-(Diethylcarbamoyl)benzeneboronic acid
Punto de fusión 138°C to 142°C
Cantidad 1 g
Formula Weight (peso de la fórmula) 221.06
Porcentaje de pureza 98%

RUO - Para uso exclusivo en investigación

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'