missing translation for 'onlineSavingsMsg'
Learn More

Clorhidrato de ácido 4-(aminometil)bencenoborónico, 96 %, Thermo Scientific Chemicals

Catalog Number p-7035141
Click to view available options
Cantidad:
1 g
5 g
75705-21-4
C7H11BClNO2
187.43
MFCD01632199
HUZNRXFJHYNUMV-UHFFFAOYSA-N
4-aminomethylphenylboronic acid hydrochloride, 4-aminomethyl phenylboronic acid hydrochloride, 4-aminomethylphenylboronic acid, hcl, 4-aminomethyl benzeneboronic acid hydrochloride, 4-aminomethylphenylboronic acid hcl, 4-aminomethylphenyl boronic acid hydrochloride, 4-aminomethyl phenyl boronic acid hydrochloride, 4-aminomethylbenzeneboronic acid hydrochloride, 4-aminomethylphenylboronic acid, hydrochloride
2734311
ácido [4-(aminometil)fenil]borónico;clorhidrato
Cl.NCC1=CC=C(C=C1)B(O)O
75705-21-4
C7H11BClNO2
187.43
MFCD01632199
HUZNRXFJHYNUMV-UHFFFAOYSA-N
4-aminomethylphenylboronic acid hydrochloride, 4-aminomethyl phenylboronic acid hydrochloride, 4-aminomethylphenylboronic acid, hcl, 4-aminomethyl benzeneboronic acid hydrochloride, 4-aminomethylphenylboronic acid hcl, 4-aminomethylphenyl boronic acid hydrochloride, 4-aminomethyl phenyl boronic acid hydrochloride, 4-aminomethylbenzeneboronic acid hydrochloride, 4-aminomethylphenylboronic acid, hydrochloride
2734311
ácido [4-(aminometil)fenil]borónico;clorhidrato
Cl.NCC1=CC=C(C=C1)B(O)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Notas
Almacenar en un lugar fresco y seco en un recipiente bien sellado. Almacenar lejos de agentes oxidantes.
TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 4-(Aminomethyl)benzeneboronic acid hydrochloride
Punto de fusión 240°C to 250°C
Cantidad 1 g
Formula Weight (peso de la fórmula) 187.43
Porcentaje de pureza 96%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'