missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Metoxifenoxi)anilina, 97 %, Thermo Scientific™

Catalog Number p-7031477
Click to view available options
Cantidad:
1 g
5 g
Este artículo no se puede devolver. Vea la política de devoluciones
13066-01-8
C13H13NO2
215.252
MFCD02663341
XFOFRBMGVDBINH-UHFFFAOYSA-N
4-2-methoxyphenoxy aniline, benzenamine, 4-2-methoxyphenoxy, 4-2-methoxyphenoxy phenylamine, 4-2-methoxyphenoxy-aniline, 4-2-methoxy-phenoxy-phenylamine
1121067
4-(2-metoxifenoxi)anilina
COC1=CC=CC=C1OC2=CC=C(C=C2)N
Este artículo no se puede devolver. Vea la política de devoluciones
13066-01-8
C13H13NO2
215.252
MFCD02663341
XFOFRBMGVDBINH-UHFFFAOYSA-N
4-2-methoxyphenoxy aniline, benzenamine, 4-2-methoxyphenoxy, 4-2-methoxyphenoxy phenylamine, 4-2-methoxyphenoxy-aniline, 4-2-methoxy-phenoxy-phenylamine
1121067
4-(2-metoxifenoxi)anilina
COC1=CC=CC=C1OC2=CC=C(C=C2)N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 4-(2-Methoxyphenoxy)aniline
Cantidad 1 g
Formula Weight (peso de la fórmula) 215.25
Porcentaje de pureza 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'