Learn More
2-n-Propiltiofeno, 97 %
Catalog Number p-7043847
Click to view available options
Cantidad:
5 g
25 g
1551-27-5
C7H10S
126.217
MFCD00014533
BTXIJTYYMLCUHI-UHFFFAOYSA-N
2-n-propylthiophene, thiophene, 2-propyl, isopropylthiophene, 2-n-propyl thiophene, 2-propyl-thiophene, 2-propyl thiophene, thiophene, propyl, thiophene,2-propyl, 2-propylthiophene, 4-hydroxy-benzenesulfonamide
73771
2-propiltiofeno
CCCC1=CC=CS1
1551-27-5
C7H10S
126.217
MFCD00014533
BTXIJTYYMLCUHI-UHFFFAOYSA-N
2-n-propylthiophene, thiophene, 2-propyl, isopropylthiophene, 2-n-propyl thiophene, 2-propyl-thiophene, 2-propyl thiophene, thiophene, propyl, thiophene,2-propyl, 2-propylthiophene, 4-hydroxy-benzenesulfonamide
73771
2-propiltiofeno
CCCC1=CC=CS1
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 2-n-Propylthiophene
Densidad 0.984
Punto de ebullición 158°C to 160°C
Temperatura de inflamación 32°C (90°F)
Olor Characteristic
Índice de refracción 1.505
Cantidad 5 g
Número UN UN1993
Beilstein 105753
Formula Weight (peso de la fórmula) 126.22
Porcentaje de pureza 97%