missing translation for 'onlineSavingsMsg'
Learn More

Éster de pinacol de ácido 2-cloro-3-cianopiridina-4-borónico, 95 %, Thermo Scientific™

Catalog Number p-7029781
Click to view available options
Cantidad:
250 mg
1 g
Este artículo no se puede devolver. Vea la política de devoluciones
878194-94-6
C12H14BClN2O2
264.516
MFCD09037479
YYPZHORJYDCOSQ-UHFFFAOYSA-N
2-chloro-3-cyanopyridine-4-boronic acid pinacol ester, 2-chloro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl nicotinonitrile, 2-chloro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine-3-carbonitrile, 2-chloro-3-cyano-4-pyridinylboronoic acid pinacol, 2-chloro-3-cyano-4-pyridinylboronic acid pinacol ester, 2-chloro-3-cyano-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-chloro-4-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine-3-carbonitrile
44755174
2-cloro-4-(4,4,5,5-tetrametil-1,3,2-dioxaborolan-2-il)piridina-3-carbonitrilo
B1(OC(C(O1)(C)C)(C)C)C2=C(C(=NC=C2)Cl)C#N
Este artículo no se puede devolver. Vea la política de devoluciones
878194-94-6
C12H14BClN2O2
264.516
MFCD09037479
YYPZHORJYDCOSQ-UHFFFAOYSA-N
2-chloro-3-cyanopyridine-4-boronic acid pinacol ester, 2-chloro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl nicotinonitrile, 2-chloro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine-3-carbonitrile, 2-chloro-3-cyano-4-pyridinylboronoic acid pinacol, 2-chloro-3-cyano-4-pyridinylboronic acid pinacol ester, 2-chloro-3-cyano-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-chloro-4-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine-3-carbonitrile
44755174
2-cloro-4-(4,4,5,5-tetrametil-1,3,2-dioxaborolan-2-il)piridina-3-carbonitrilo
B1(OC(C(O1)(C)C)(C)C)C2=C(C(=NC=C2)Cl)C#N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 2-Chloro-3-cyanopyridine-4-boronic acid pinacol ester
Punto de fusión 100°C to 101°C
Cantidad 250 mg
Formula Weight (peso de la fórmula) 264.52
Porcentaje de pureza 95%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'